Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCTGTCCCCTCAGCCGCTCCAG[C/T]TTCCTCCGAAGGTCTGGGGGCAACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cpxm1 PubMed Links |
Cpxm1 - carboxypeptidase X 1 (M14 family) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019696.2 | 2265 | Silent Mutation | NP_062670.2 |