Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATATCCCCTCCCACCGCCACT[C/T]ACCTAGGTTGACGAAGCTCATGACC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bmp7 PubMed Links |
Bmp7 - bone morphogenetic protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007557.3 | Intron | NP_031583.2 |