Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGACAAGGGGTTACATAGACA[A/C]TGGTCCGCTTGGCAGAAGGGGAAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bahcc1 PubMed Links |
Bahcc1 - BAH domain and coiled-coil containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198423.3 | Intron | NP_940815.3 |