Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Foxk2 PubMed Links |
Foxk2 - forkhead box K2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Narf - nuclear prelamin A recognition factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026272.3 | 3795 | UTR 3 | NP_080548.3 |