Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAATATCATAGGCCAGATCACA[A/G]GGGACAGATTGTAAAGTATTCTAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nrxn3 PubMed Links |
Nrxn3 - neurexin III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198587.3 | Intron | NP_001185516.2 | ||||
NM_001252074.2 | Intron | NP_001239003.1 | ||||
NM_172544.3 | Intron | NP_766132.2 |