Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGCTAGATTCTTTTTTCTCAG[A/T]CTAATAGACATGTTGGGAAACATCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Iqgap2 PubMed Links |
Iqgap2 - IQ motif containing GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027711.1 | Intron | NP_081987.1 |