Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGCATCTGTGAGAGTGTGACAG[C/T]GCAGGGACGGGACTAGGACATGGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Epas1 PubMed Links |
Epas1 - endothelial PAS domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010137.3 | Intron | NP_034267.3 |