Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTACTTAGTCAACCCTTTCCTAG[C/A]CACCGTAAGAGTCTAATAATAAGGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Adcyap1 PubMed Links |
Adcyap1 - adenylate cyclase activating polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009625.2 | 197 | UTR 5 | NP_033755.1 |