Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-139 |
---|---|
miRBase Accession Number: | MI0002768 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UCUACAGUGCACGUGUCU |
Species: | Lagothrix lagotricha, Pan paniscus |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000117, mir-139 |
Mature miRNA Sequence: | UCUACAGUGCACGUGUCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Lagothrix lagotricha | lla-miR-139 | MIMAT0002472 |
Pan paniscus | ppa-miR-139 | MIMAT0002473 |