Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-33b |
---|---|
miRBase Accession Number: | MI0003646 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GUGCAUUGCUGUUGCAUUGC |
Species: | Human, Bovine, Dog, Horse, Pan troglodytes, Pig, Pteropus alecto, Rhesus monkey |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000070, mir-33 |
Mature miRNA Sequence: | GUGCAUUGCUGUUGCAUUGC |
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-33b-5p | MIMAT0003301 | |
Bovine | bta-miR-33b | MIMAT0009295 | |
Dog | cfa-miR-33b | MIMAT0009862 | |
Horse | eca-miR-33b | MIMAT0013037 | |
Pan troglodytes | ptr-miR-33b | MIMAT0008108 | |
Pig | ssc-miR-33b-5p | MIMAT0044654 | |
View More Rows | |||
Pteropus alecto | pal-miR-33b-5p | MIMAT0040300 | |
Rhesus monkey | mml-miR-33b-5p | MIMAT0006173 |