Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-221-5p |
---|---|
Stem-loop Accession Number: | MI0000298 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACCUGGCAUACAAUGUAGAUUU |
Species: | Human, Alligator mississippiensis, Columba livia, Paralichthys olivaceus |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000051, mir-221 |
Mature miRNA Sequence: | ACCUGGCAUACAAUGUAGAUUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-221-5p | MIMAT0004568 |
Alligator mississippiensis | ami-miR-221-5p | MIMAT0038252 |
Columba livia | cli-miR-221-5p | MIMAT0038604 |
Paralichthys olivaceus | pol-miR-221-5p | MIMAT0025448 |