Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | eca-miR-9023 |
---|---|
miRBase Accession Number: | MI0028187 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUGUAUUAAUACGGUAGGUAGUG |
Species: | Horse |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUGUAUUAAUACGGUAGGUAGUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Horse | eca-miR-9023 | MIMAT0034565 |