Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101620.3 | NP_173201.2 | 32 - 33 | 4196 | 67 | |
GenBank mRNA | Z28389.1 | - | 4143 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |