Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G18070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035986.1 | NP_001031063.1 | 6 - 7 | 456 | 125 | |
NM_001198097.1 | NP_001185026.1 | 6 - 7 | 461 | 125 | ||
NM_101669.3 | NP_173247.1 | 6 - 7 | 505 | 125 | ||
GenBank mRNA | AY120769.1 | - | 6 - 7 | 440 | 125 | |
BT008889.1 | - | 5 - 6 | 332 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |