Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126178.4 | NP_565254.1 | 7 - 8 | 1312 | 71 | |
NM_201658.1 | NP_973387.1 | 6 - 7 | 1321 | 71 | ||
GenBank mRNA | AF019637.1 | - | 7 - 8 | 1231 | 71 | |
BT020534.1 | - | 6 - 7 | 999 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |