Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337357.1 | NP_001326912.1 | 3 - 4 | 1424 | 64 | |
NM_111043.3 | NP_566151.1 | 3 - 4 | 1435 | 64 | ||
GenBank mRNA | AY057662.1 | - | 3 - 4 | 1373 | 64 | |
AY099729.1 | - | 3 - 4 | 1380 | 64 | ||
AY128906.1 | - | 2 - 3 | 1125 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |