Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341092.1 | NP_001319956.1 | 4 - 5 | 982 | 66 | |
NM_117725.3 | NP_193363.4 | 4 - 5 | 569 | 66 | ||
NM_148347.2 | NP_680713.2 | 5 - 6 | 778 | 66 | ||
NM_179211.3 | NP_849542.2 | 3 - 4 | 3213 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |