Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102887.5 | NP_174433.2 | 10 - 11 | 1474 | 126 | |
GenBank mRNA | AB073133.1 | - | 10 - 11 | 1324 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |