Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102940.6 | NP_174486.1 | 4 - 5 | 1403 | 95 | |
GenBank mRNA | AY044335.1 | - | 4 - 5 | 934 | 95 | |
AY084576.1 | - | 4 - 5 | 1043 | 95 | ||
AY128355.1 | - | 4 - 5 | 1040 | 95 | ||
BT000019.1 | - | 4 - 5 | 934 | 95 | ||
X58149.1 | - | 1012 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |