Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127114.2 | NP_565375.1 | 1 - 2 | 125 | 96 | |
GenBank mRNA | DQ446500.1 | - | 1 - 2 | 66 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |