Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337993.1 | NP_001319530.1 | 3 - 4 | 375 | 67 | |
NM_180239.4 | NP_850570.2 | 2 - 3 | 464 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |