Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204039.1 | NP_001190968.1 | 11 - 12 | 923 | 112 | |
NM_001342543.1 | NP_001328265.1 | 11 - 12 | 987 | 112 | ||
NM_120106.3 | NP_568060.1 | 11 - 12 | 987 | 112 | ||
GenBank mRNA | AJ627908.1 | - | 10 - 11 | 770 | 112 | |
AM260490.1 | - | 10 - 11 | 770 | 112 | ||
AY046052.1 | - | 11 - 12 | 854 | 112 | ||
AY079349.1 | - | 10 - 11 | 770 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |