Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120241.4 | NP_195783.3 | 12 - 13 | 2557 | 111 | |
GenBank mRNA | AJ488305.1 | - | 12 - 13 | 2362 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |