Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342651.1 | NP_001318457.1 | 13 - 14 | 5954 | 143 | |
NM_001342652.1 | NP_001331954.1 | 13 - 14 | 5977 | 143 | ||
NM_120309.2 | NP_195851.2 | 12 - 13 | 5619 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |