Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198152.1 | NP_001185081.1 | 3 - 4 | 587 | 90 | |
NM_102272.4 | NP_564214.2 | 3 - 4 | 580 | 90 | ||
NM_180622.3 | NP_850953.1 | 3 - 4 | 602 | 90 | ||
GenBank mRNA | AF015552.1 | - | 3 - 4 | 350 | 90 | |
AY088272.1 | - | 3 - 4 | 371 | 90 | ||
BT033157.1 | - | 3 - 4 | 344 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |