Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203301.2 | NP_001190230.1 | 4 - 5 | 1008 | 65 | |
NM_120640.4 | NP_196177.1 | 4 - 5 | 986 | 65 | ||
GenBank mRNA | AF361837.1 | - | 4 - 5 | 901 | 65 | |
AY078043.1 | - | 4 - 5 | 717 | 65 | ||
D17578.1 | - | 4 - 5 | 854 | 65 | ||
L27158.1 | - | 4 - 5 | 777 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |