Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127736.4 | NP_179759.1 | 1 - 2 | 424 | 60 | |
GenBank mRNA | AK229516.1 | - | 1 - 2 | 289 | 60 | |
BT008554.1 | - | 1 - 2 | 281 | 60 | ||
BT008698.1 | - | 1 - 2 | 289 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |