Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127812.3 | NP_565537.1 | 1 | 636 | 118 | |
GenBank mRNA | AY062726.1 | - | 1 | 552 | 118 | |
AY093356.1 | - | 1 | 468 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |