Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345460.1 | NP_001330163.1 | 8 - 9 | 1334 | 83 | |
NM_125518.5 | NP_200933.2 | 9 - 10 | 1393 | 83 | ||
NM_180907.3 | NP_851238.1 | 9 - 10 | 1395 | 83 | ||
GenBank mRNA | AK317449.1 | - | 9 - 10 | 1385 | 83 | |
AY099736.1 | - | 9 - 10 | 1316 | 83 | ||
BT002603.1 | - | 8 - 9 | 1235 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |