Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125893.5 | NP_201299.2 | 12 - 13 | 1757 | 75 | |
NM_180940.2 | NP_851271.1 | 12 - 13 | 1757 | 75 | ||
GenBank mRNA | AY059877.1 | - | 12 - 13 | 1740 | 75 | |
AY064044.1 | - | 12 - 13 | 1716 | 75 | ||
AY096382.1 | - | 12 - 13 | 1595 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |