Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115410.3 | NP_191112.1 | 7 - 8 | 741 | 87 | |
GenBank mRNA | AF386995.1 | - | 7 - 8 | 711 | 87 | |
AK317055.1 | - | 7 - 8 | 728 | 87 | ||
BT006305.1 | - | 6 - 7 | 573 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |