Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125287.5 | NP_568894.1 | 2 - 3 | 577 | 70 | |
NM_203233.1 | NP_974962.1 | 2 - 3 | 543 | 70 | ||
GenBank mRNA | AB021706.1 | - | 2 - 3 | 318 | 70 | |
JQ937241.1 | - | 2 - 3 | 303 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |