Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115977.4 | NP_191672.1 | 4 - 5 | 636 | 77 | |
GenBank mRNA | DQ370437.1 | - | 3 - 4 | 234 | 77 | |
KJ138782.1 | - | 3 - 4 | 234 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |