Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128552.4 | NP_180559.1 | 7 - 8 | 1211 | 87 | |
NM_179808.2 | NP_850139.1 | 6 - 7 | 1136 | 87 | ||
GenBank mRNA | AY063966.1 | - | 7 - 8 | 1073 | 87 | |
AY096462.1 | - | 7 - 8 | 967 | 87 | ||
D17579.1 | - | 7 - 8 | 1049 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |