Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GASA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085118.2 | NP_001078587.1 | 1 - 2 | 256 | 74 | |
NM_121527.4 | NP_197027.1 | 2 - 3 | 302 | 74 | ||
GenBank mRNA | AF360199.1 | - | 2 - 3 | 215 | 74 | |
AY040048.1 | - | 2 - 3 | 101 | 74 | ||
AY057601.1 | - | 2 - 3 | 215 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |