Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGSIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121853.4 | NP_197349.2 | 7 - 8 | 1540 | 136 | |
GenBank mRNA | AK118951.1 | - | 7 - 8 | 1381 | 136 | |
KJ138680.1 | - | 7 - 8 | 1345 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |