Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKdZeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128652.4 | NP_180655.1 | 5 - 6 | 714 | 100 | |
GenBank mRNA | AY064020.1 | - | 5 - 6 | 664 | 100 | |
AY094423.1 | - | 5 - 6 | 690 | 100 | ||
AY096698.1 | - | 5 - 6 | 611 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |