Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340129.1 | NP_001327870.1 | 4 - 5 | 645 | 87 | |
NM_116048.3 | NP_567119.1 | 5 - 6 | 753 | 87 | ||
GenBank mRNA | AF334717.1 | - | 5 - 6 | 471 | 87 | |
AY059746.1 | - | 5 - 6 | 691 | 87 | ||
AY070463.1 | - | 5 - 6 | 756 | 87 | ||
AY091392.1 | - | 5 - 6 | 471 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |