Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340324.1 | NP_001328724.1 | 8 - 9 | 605 | 92 | |
NM_001340325.1 | NP_001328720.1 | 7 - 8 | 693 | 92 | ||
NM_001340326.1 | NP_001328725.1 | 7 - 8 | 750 | 92 | ||
NM_001340327.1 | NP_001328721.1 | 7 - 8 | 678 | 92 | ||
NM_001340328.1 | NP_001328726.1 | 6 - 7 | 528 | 92 | ||
NM_001340329.1 | NP_001328727.1 | 4 - 5 | 490 | 92 | ||
NM_116381.4 | NP_192060.2 | 8 - 9 | 616 | 92 | ||
GenBank mRNA | AY758071.1 | - | 8 - 9 | 597 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |