Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036498.3 | NP_001031575.1 | 13 - 14 | 1330 | 87 | |
NM_001036499.1 | NP_001031576.1 | 13 - 14 | 1266 | 87 | ||
NM_001203732.1 | NP_001190661.1 | 13 - 14 | 1328 | 87 | ||
NM_116491.3 | NP_567243.1 | 12 - 13 | 1424 | 87 | ||
GenBank mRNA | AJ318017.1 | - | 11 - 12 | 1125 | 87 | |
AK317123.1 | - | 13 - 14 | 1267 | 87 | ||
AY046030.1 | - | 12 - 13 | 1347 | 87 | ||
AY133878.1 | - | 11 - 12 | 1091 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |