Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116659.5 | NP_567262.1 | 3 - 4 | 1050 | 69 | |
GenBank mRNA | AY085333.1 | - | 3 - 4 | 555 | 69 | |
AY091099.1 | - | 3 - 4 | 540 | 69 | ||
AY122960.1 | - | 3 - 4 | 514 | 69 | ||
AY572784.1 | - | 3 - 4 | 514 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |