Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116790.3 | NP_567299.2 | 2 - 3 | 289 | 61 | |
NM_202788.1 | NP_974517.1 | 2 - 3 | 241 | 61 | ||
GenBank mRNA | BT033036.1 | - | 2 - 3 | 170 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |