Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116790.3 | NP_567299.2 | 8 - 9 | 684 | 85 | |
NM_202788.1 | NP_974517.1 | 8 - 9 | 633 | 85 | ||
GenBank mRNA | BT033036.1 | - | 8 - 9 | 565 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |