Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116964.3 | NP_192634.1 | 1 | 1128 | 186 | |
GenBank mRNA | AY054486.1 | - | 1 | 1041 | 186 | |
AY093295.1 | - | 1 | 969 | 186 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |