Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116971.6 | NP_192641.2 | 8 - 9 | 1092 | 99 | |
GenBank mRNA | AK227049.1 | - | 8 - 9 | 929 | 99 | |
AY091058.1 | - | 8 - 9 | 925 | 99 | ||
AY133739.1 | - | 8 - 9 | 902 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |