Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036098.2 | NP_001031175.1 | 1 - 2 | 275 | 108 | |
NM_001198275.1 | NP_001185204.1 | 1 - 2 | 283 | 108 | ||
NM_104118.3 | NP_175649.1 | 1 - 2 | 437 | 108 | ||
GenBank mRNA | AY039855.1 | - | 1 - 2 | 252 | 108 | |
AY056415.1 | - | 1 - 2 | 252 | 108 | ||
BT000515.1 | - | 1 - 2 | 176 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |