Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336552.1 | NP_001323587.1 | 2 - 3 | 183 | 111 | |
NM_129086.4 | NP_181077.3 | 6 - 7 | 743 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |