Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129089.2 | NP_181080.1 | 1 - 2 | 313 | 113 | |
GenBank mRNA | AK228462.1 | - | 1 - 2 | 180 | 113 | |
AY050446.1 | - | 1 - 2 | 209 | 113 | ||
AY058854.1 | - | 1 - 2 | 159 | 113 | ||
AY086345.1 | - | 1 - 2 | 252 | 113 | ||
AY097349.1 | - | 1 - 2 | 115 | 113 | ||
M82921.1 | - | 1 - 2 | 136 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |