Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105090.4 | NP_176599.2 | 2 - 3 | 1412 | 97 | |
GenBank mRNA | DQ499021.1 | - | 2 - 3 | 1224 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |