Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337323.1 | NP_001319439.1 | 5 - 6 | 962 | 106 | |
NM_001337324.1 | NP_001327637.1 | 4 - 5 | 860 | 106 | ||
NM_001337325.1 | NP_001327636.1 | 5 - 6 | 916 | 106 | ||
NM_110980.4 | NP_186764.1 | 4 - 5 | 860 | 106 | ||
GenBank mRNA | AK316897.1 | - | 4 - 5 | 708 | 106 | |
AY093218.1 | - | 5 - 6 | 823 | 106 | ||
BT001239.1 | - | 3 - 4 | 249 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |